Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1-methyl-1H-indole 95%

Supplier: AMBEED, INC
Warning

A210542-1G A210542-10G A210542-1G A210542-5G
103818-014EA 9 USD InStock
103818-014 103848-710 76770-500 77087-084 77087-086
5-Bromo-1-methyl-1H-indole 95%
5-Bromo-1-methyl-1H-indole
Precaución: for Research Use Only


Formula: C₉H₈BrN
MW: 210.071 g/mol
Boiling Pt: 300.9 °C (760 mm Hg)
Melting Pt: 43 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00159857
CAS Number: 10075-52-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty