Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromoindan-1-ol 97%

Supplier: AMBEED, INC
Warning

A132721-1G A132721-100MG A132721-250MG A132721-5G A132721-1G
103801-776EA 98 USD InStock
103801-776 76870-814 76870-816 77298-578 77314-362
5-Bromoindan-1-ol 97%
5-Bromoindan-1-ol
Precaución: for Research Use Only


Formula: C₉H₉BrO
Boiling Pt: 307.734 °C (760 mm Hg)
Flash Pt: 139.913 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06808667
CAS Number: 34598-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty