Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,3-dimethoxypyrazine 95%

Supplier: AMBEED, INC
Warning

A136365-1G A136365-100MG A136365-250MG
103802-294EA 78 USD InStock
103802-294 76972-358 76972-360
5-Bromo-2,3-dimethoxypyrazine 95%
5-Bromo-2,3-dimethoxypyrazine
Precaución: for Research Use Only


Formula: C₆H₇BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18205982
CAS Number: 89466-19-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty