Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,3-dimethylindole 97%

Supplier: AMBEED, INC
Warning

A278548-1G A278548-5G A278548-100MG A278548-10G A278548-250MG A278548-1G
103813-418EA 32 USD InStock
103813-418 103853-360 76977-344 76977-346 76977-348 77305-298
5-Bromo-2,3-dimethylindole 97%
5-Bromo-2,3-dimethylindole
Precaución: for Research Use Only


Formula: C₁₀H₁₀BrN
Boiling Pt: 340.35 °C (760 mm Hg)
Melting Pt: 136…137 °C
Flash Pt: 159.638 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00850844
CAS Number: 4583-55-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty