Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,4-dimethylpyrimidine 98%

Supplier: AMBEED, INC
Warning

A111325-1G A111325-100MG A111325-250MG A111325-1G
103797-818EA 350 USD InStock
103797-818 103823-634 76821-850 76821-852 77310-438
5-Bromo-2,4-dimethylpyrimidine 98%
5-Bromo-2,4-dimethylpyrimidine
Precaución: for Research Use Only


Formula: C₆H₇BrN₂
MW: 187.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13705449
CAS Number: 69696-37-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty