Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-6-chloro-2-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A189848-1G A189848-5G A189848-100G A189848-25G A189848-1G
103816-226EA 5 USD InStock
103816-226 103846-822 77012-254 77084-592 77302-188
3-Bromo-6-chloro-2-methylpyridine 98%
3-Bromo-6-chloro-2-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅BrClN
MW: 206.47
Boiling Pt: 399 °C (760 mm Hg)
Melting Pt: 130 °C
Flash Pt: 87.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07375117
CAS Number: 132606-40-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty