Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-fluoronicotinaldehyde 98%

Supplier: AMBEED, INC
Warning

A244811-1G A244811-5G A244811-10G A244811-1G A244811-25G A244811-5G A244811-100G
103811-602EA 13 USD InStock
103811-602 103851-436 76786-652 77090-650 77090-652 77090-654 77872-225
5-Bromo-2-fluoronicotinaldehyde 98%
5-Bromo-2-fluoronicotinaldehyde
Precaución: for Research Use Only


Formula: C₆H₃BrFNO
Boiling Pt: 258.2 °C (760 mm Hg)
Melting Pt: 86…90 °C
Flash Pt: 109.936 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09842440
CAS Number: 875781-15-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty