Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-fluorophenylacetic acid 98%

Supplier: AMBEED, INC
Warning

A720993-1G A720993-25G A720993-5G A720993-100G A720993-10G
103840-744EA 13 USD InStock
103840-744 76844-956 76844-958 76844-960 77243-652 77243-654
5-Bromo-2-fluorophenylacetic acid 98%
5-Bromo-2-fluorophenylacetic acid
Precaución: for Research Use Only


Formula: C₈H₆BrFO₂
Boiling Pt: 319.6 °C (760 mm Hg)
Melting Pt: 111…112 °C
Storage Temperature: Ambient
MDL Number: MFCD06246020
CAS Number: 883514-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty