Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methoxynicotinonitrile 97%

Supplier: AMBEED, INC
Warning

A380944-1G A380944-5G A380944-10G
103826-686EA 24 USD InStock
103826-686 103837-720 76841-038 77872-597
5-Bromo-2-methoxynicotinonitrile 97%
5-Bromo-2-methoxynicotinonitrile
Precaución: for Research Use Only


Formula: C₇H₅BrN₂O
MW: 213.031 g/mol
Boiling Pt: 266.5 °C (760 mm Hg)
Melting Pt: 110…115 °C
Flash Pt: 115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09801047
CAS Number: 941294-54-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty