Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methyl-2H-pyrazolo[3,4-b]pyridine 95%

Supplier: AMBEED, INC
Warning

A395291-5G A395291-1G A395291-10G A395291-250MG
103826-960EA 58 USD InStock
103826-960 103837-998 76841-418 77155-562 77155-564
5-Bromo-2-methyl-2H-pyrazolo[3,4-b]pyridine 95%
5-Bromo-2-methyl-2H-pyrazolo[3,4-b]pyridine
Precaución: for Research Use Only


Formula: C₇H₆BrN₃
Storage Temperature: Ambient
MDL Number: MFCD19443914
CAS Number: 1316852-65-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty