Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-o-toluidine 97%

Supplier: AMBEED, INC
Warning

A109306-5G A109306-1000G A109306-100G A109306-10G A109306-25G A109306-500G A109306-5G
103823-284EA 17 USD InStock
103823-284 76970-634 76970-636 76970-638 76970-640 76970-642 77310-102
5-Bromo-o-toluidine 97%
5-Bromo-o-toluidine
Precaución: for Research Use Only


Formula: C₇H₈BrN
Boiling Pt: 260.3 °C (760 mm Hg)
Melting Pt: 33 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00800678
CAS Number: 39478-78-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty