Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-o-toluic acid 98%

Supplier: AMBEED, INC
Warning

A363133-25G A363133-5G A363133-100G A363133-10G A363133-1G A363133-500G
103837-352EA 6 USD InStock
103837-352 76840-572 76840-574 77240-138 77240-140 77240-142 77240-144
5-Bromo-o-toluic acid 98%
5-Bromo-o-toluic acid


Formula: C₈H₇BrO₂
Boiling Pt: 319.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00267350
CAS Number: 79669-49-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty