Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methylbenzothiazole 97%

Supplier: AMBEED, INC
Warning

A239924-1G A239924-5G A239924-10G A239924-250MG A239924-1G A239924-5G A239924-25G A239924-100G
103811-318EA 13 USD InStock
103811-318 103851-116 76771-386 76771-388 77090-268 77090-270 77304-176 77877-799
5-Bromo-2-methylbenzothiazole 97%
5-Bromo-2-methylbenzothiazole
Precaución: for Research Use Only


Formula: C₈H₆BrNS
Melting Pt: 75…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00603096
CAS Number: 63837-11-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty