Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-phenyloxazole 97%

Supplier: AMBEED, INC
Warning

A221453-1G A221453-5G A221453-100MG A221453-250MG A221453-1G A221453-5G
103818-926EA 83 USD InStock
103818-926 103849-806 76770-824 76770-826 77303-600 77303-602
5-Bromo-2-phenyloxazole 97%
5-Bromo-2-phenyloxazole
Precaución: for Research Use Only


Formula: C₉H₆BrNO
Boiling Pt: 308.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00466256
CAS Number: 92629-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty