Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-(tert-butyl)-2-hydroxybenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A424828-1G A424828-5G A424828-100MG A424828-10G A424828-250MG
103827-450EA 62 USD InStock
103827-450 103838-498 76842-148 76842-150 77240-842 77240-844 77240-846
5-Bromo-3-(tert-butyl)-2-hydroxybenzaldehyde 95%
5-Bromo-3-(tert-butyl)-2-hydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₁₃BrO₂
MW: 257.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00456060
CAS Number: 153759-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty