Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-fluoropicolinonitrile 98%

Supplier: AMBEED, INC
Warning

A122326-1G A122326-10G A122326-250MG A122326-25G A122326-5G
103800-204EA 9 USD InStock
103800-204 77009-470 77009-472 77009-474 77312-342
5-Bromo-3-fluoropicolinonitrile 98%
5-Bromo-3-fluoropicolinonitrile
Precaución: For research use only.


Formula: C₆H₂BrFN₂
MW: 200.9959 g/mol
Boiling Pt: 238 °C (760 mm Hg)
Melting Pt: 103…105 °C
Flash Pt: 97.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07375030
CAS Number: 886373-28-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty