Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromo-3-methoxypyridine 98%

Supplier: AMBEED, INC
Warning

A240596-1G A240596-5G A240596-100G A240596-100MG A240596-10G A240596-250MG A240596-1G A240596-25G A240596-5G
103811-352EA 9 USD InStock
103811-352 103851-154 77000-412 77000-414 77000-416 77000-418 77090-308 77090-310 77090-312
2-Amino-5-bromo-3-methoxypyridine 98%
2-Amino-5-bromo-3-methoxypyridine
Precaución: for Research Use Only


Formula: C₆H₇BrN₂O
MW: 203.04 g/mol
Boiling Pt: 253.7 °C (760 mm Hg)
Flash Pt: 107.255 °C
Storage Temperature: Ambient
MDL Number: MFCD09909849
CAS Number: 42409-58-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty