Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-methylindole 97%

Supplier: AMBEED, INC
Warning

A258902-5G A258902-250MG A258902-1G A258902-5G A258902-25G
103812-408EA 50 USD InStock
103812-408 103852-286 76771-826 77091-820 77091-822 77304-756
5-Bromo-3-methylindole 97%
5-Bromo-3-methylindole
Precaución: For research use only.


Formula: C₉H₈BrN
MW: 210.08 g/mol
Boiling Pt: 325.1 °C (760 mm Hg)
Flash Pt: 150.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038746
CAS Number: 10075-48-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty