Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-methylpyrazin-2-ol 97%

Supplier: AMBEED, INC
Warning

A535967-1G A535967-100MG A535967-250MG A535967-5G A535967-1G
103828-592EA 37 USD InStock
103828-592 76991-558 76991-560 76991-562 77307-758
5-Bromo-3-methylpyrazin-2-ol 97%
5-Bromo-3-methylpyrazin-2-ol
Precaución: for Research Use Only


Formula: C₅H₅BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD22548397
CAS Number: 100047-56-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty