Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-hydroxypyridine 98%

Supplier: AMBEED, INC
Warning

A241929-1G A241929-100G A241929-10G A241929-500G A241929-5G A241929-1G A241929-25G
103811-444EA 9 USD InStock
103811-444 103851-252 76987-638 76987-640 76987-642 77090-416 77304-230 77304-232
3-Bromo-5-hydroxypyridine 98%
3-Bromo-5-hydroxypyridine
Precaución: for Research Use Only


Formula: C₅H₄BrNO
MW: 174
Melting Pt: 166…170 °C
Flash Pt: 161.652 °C
Storage Temperature: Ambient
MDL Number: MFCD00661305
CAS Number: 74115-13-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty