Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-chloro-2-aminopyridine 97%

Supplier: AMBEED, INC
Warning

A116738-1G A116738-10G A116738-250MG A116738-25G A116738-1G A116738-5G
103798-790EA 6 USD InStock
103798-790 103824-658 76781-424 76781-426 76781-428 77297-494 77311-454
5-Bromo-4-chloro-2-aminopyridine 97%
5-Bromo-4-chloro-2-aminopyridine
Precaución: for Research Use Only


Formula: C₅H₄BrClN₂
Boiling Pt: 271.5 °C (760 mm Hg)
Flash Pt: 118.038 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12407282
CAS Number: 942947-94-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty