Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-fluoro-2-hydroxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A152062-1G A152062-5G A152062-250MG A152062-1G A152062-5G
103804-346EA 17 USD InStock
103804-346 103842-500 76783-406 77318-106 77318-108
5-Bromo-4-fluoro-2-hydroxybenzaldehyde 97%
5-Bromo-4-fluoro-2-hydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄BrFO₂
Boiling Pt: 256.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06739493
CAS Number: 399-00-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty