Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-chloro-1H-indole 95%

Supplier: AMBEED, INC
Warning

A111477-1G A111477-5G A111477-100G A111477-100MG A111477-10G A111477-250MG A111477-25G A111477-1G A111477-5G
103797-842EA 12 USD InStock
103797-842 103823-662 76869-730 76869-732 76869-734 76869-736 76869-738 77284-122 77310-464
5-Bromo-6-chloro-1H-indole 95%
5-Bromo-6-chloro-1H-indole
Precaución: for Research Use Only


Formula: C₈H₅BrClN
MW: 230.48 g/mol
Boiling Pt: 358 °C (760 mm Hg)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD11848566
CAS Number: 122531-09-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty