Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-chloronicotinic acid 97%

Supplier: AMBEED, INC
Warning

A383409-1G A383409-25G A383409-5G A383409-100G A383409-10G A383409-250MG A383409-500G
103826-740EA 6 USD InStock
103826-740 103837-776 76841-104 76841-106 76841-108 77003-654 77003-656 77003-658 77672-898
5-Bromo-6-chloronicotinic acid 97%
5-Bromo-6-chloronicotinic acid
Precaución: for Research Use Only


Formula: C₆H₃BrClNO₂
Boiling Pt: 370 °C (760 mm Hg)
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD01927098
CAS Number: 29241-62-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty