Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-chloropicolinonitrile 97%

Supplier: AMBEED, INC
Warning

A233615-1G A233615-5G A233615-100MG A233615-250MG
103810-894EA 167 USD InStock
103810-894 103850-656 77013-576 77013-578
5-Bromo-6-chloropicolinonitrile 97%
5-Bromo-6-chloropicolinonitrile
Precaución: for Research Use Only


Formula: C₆H₂BrClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD18257828
CAS Number: 1256823-65-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty