Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-methoxypicolinic acid 97%

Supplier: AMBEED, INC
Warning

A220611-1G A220611-5G A220611-100MG A220611-250MG A220611-1G A220611-5G
103818-862EA 39 USD InStock
103818-862 103849-744 76826-178 76826-180 77088-332 77088-334
5-Bromo-6-methoxypicolinic acid 97%
5-Bromo-6-methoxypicolinic acid
Precaución: for Research Use Only


Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185810
CAS Number: 1214334-70-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty