Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-7-fluoro-2,3-dihydro-1H-inden-1-one 95%

Supplier: AMBEED, INC
Warning

A651351-100MG A651351-10G A651351-250MG A651351-1G A651351-25G A651351-5G
103840-400EA 100 USD InStock
103840-400 76819-742 76819-744 76819-746 76844-570 76844-572 76844-574
5-Bromo-7-fluoro-2,3-dihydro-1H-inden-1-one 95%
5-Bromo-7-fluoro-2,3-dihydro-1H-inden-1-one
Precaución: for Research Use Only


Formula: C₉H₆BrFO
MW: 229.05 g/mol
Boiling Pt: 311.0±42.0 °C (760 mm Hg)
Melting Pt: 135…139 °C
Flash Pt: 141.9 °C
Storage Temperature: Ambient
MDL Number: MFCD17677343
CAS Number: 1242157-14-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty