Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-8-fluoroisoquinoline 98%

Supplier: AMBEED, INC
Warning

A140470-1G A140470-100MG A140470-250MG A140470-1G
103802-870EA 139 USD InStock
103802-870 76823-410 76823-412 77299-148
5-Bromo-8-fluoroisoquinoline 98%
5-Bromo-8-fluoroisoquinoline
Precaución: for Research Use Only


Formula: C₉H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD18447885
CAS Number: 679433-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty