Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromobenzene-1,2,3-triol 95%

Supplier: AMBEED, INC
Warning

A586436-1G A586436-5G A586436-100MG A586436-250MG
103828-890EA 66 USD InStock
103828-890 103840-038 76844-178 76844-180 77157-478 77157-480
5-Bromobenzene-1,2,3-triol 95%
5-Bromobenzene-1,2,3-triol
Precaución: for Research Use Only


Formula: C₆H₅BrO₃
MW: 205.006 g/mol
Boiling Pt: 345.9 °C (760 mm Hg)
Melting Pt: 118 °C
Flash Pt: 163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22555992
CAS Number: 16492-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty