Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromobenzofuran-2-carboxylic acid 96%

Supplier: AMBEED, INC
Warning

A591958-1G A591958-5G A591958-250MG
103828-932EA 29 USD InStock
103828-932 103840-078 76844-226 76844-228 77266-650
5-Bromobenzofuran-2-carboxylic acid 96%
5-Bromobenzofuran-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₅BrO₃
Boiling Pt: 370.9 °C (760 mm Hg)
Melting Pt: 257 °C (760 mm Hg)
Flash Pt: 178.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00093973
CAS Number: 10242-11-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty