Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-N1-methylbenzene-1,2-diamine 96%

Supplier: AMBEED, INC
Warning

A441832-1G A441832-5G A441832-10G A441832-250MG A441832-25G
103827-678EA 35 USD InStock
103827-678 103838-748 76842-516 76842-518 77034-022 77034-024 77034-026
5-Bromo-N1-methylbenzene-1,2-diamine 96%
5-Bromo-N1-methylbenzene-1,2-diamine
Precaución: for Research Use Only


Formula: C₇H₉BrN₂
Boiling Pt: 289 °C (760 mm Hg)
Flash Pt: 129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11977373
CAS Number: 337915-79-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty