Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromopentyl acetate 95%

Supplier: AMBEED, INC
Warning

A734085-100G A734085-25G A734085-5G
103840-798EA 14 USD InStock
103840-798 76776-112 76845-022 76845-024
5-Bromopentyl acetate 95%
5-Bromopentyl acetate
Precaución: for Research Use Only


Formula: C₇H₁₃BrO₂
Boiling Pt: 224.1 °C (760 mm Hg)
Flash Pt: 111.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00039204
CAS Number: 15848-22-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty