Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromopicolinic acid 98%

Supplier: AMBEED, INC
Warning

A635338-25G A635338-5G A635338-100G A635338-10G A635338-500G
103840-310EA 11 USD InStock
103840-310 76844-480 76844-482 77280-438 77280-440 77280-442
5-Bromopicolinic acid 98%
5-Bromopicolinic acid
Precaución: for Research Use Only


Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 319.5 °C (760 mm Hg)
Melting Pt: 173…175 °C
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00234149
CAS Number: 30766-11-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty