Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromothiophene-3-carboxaldehyde 95%

Supplier: AMBEED, INC
Warning

A278892-1G A278892-5G A278892-250MG A278892-1G
103813-436EA 39 USD InStock
103813-436 103853-380 77032-130 77093-308
5-Bromothiophene-3-carboxaldehyde 95%
5-Bromothiophene-3-carboxaldehyde
Precaución: for Research Use Only


Formula: C₅H₃BrOS
MW: 191.046 g/mol
Boiling Pt: 237…242 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09042555
CAS Number: 18791-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty