Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2,3-difluoropyridine 96%

Supplier: AMBEED, INC
Danger

A189160-5G A189160-10G A189160-25G A189160-5G
103809-226EA 291 USD InStock
103809-226 103846-758 76986-036 77084-518 77084-520
5-Chloro-2,3-difluoropyridine 96%
5-Chloro-2,3-difluoropyridine
Precaución: for Research Use Only


Formula: C5H2ClF2N
Boiling Pt: 135…136 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04039314
CAS Number: 89402-43-7
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty