Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2,4-difluorobenzaldehyde 96%

Supplier: AMBEED, INC
Warning

A100867-1G A100867-5G A100867-250MG A100867-1G A100867-5G
103795-660EA 22 USD InStock
103795-660 103821-352 76765-138 77296-862 77296-864
5-Chloro-2,4-difluorobenzaldehyde 96%
5-Chloro-2,4-difluorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₃ClF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD07782052
CAS Number: 695187-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty