Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-1-fluoro-2-nitrobenzene 97%

Supplier: AMBEED, INC
Warning

A213047-5G A213047-25G A213047-100G A213047-10G A213047-500G A213047-5G
103848-958EA 5 USD InStock
103848-958 77087-438 77152-478 77152-480 77152-482 77303-268
4-Chloro-1-fluoro-2-nitrobenzene 97%
4-Chloro-1-fluoro-2-nitrobenzene
Precaución: for Research Use Only


Formula: C₆H₃ClFNO₂
Boiling Pt: 236 °C (760 mm Hg)
Melting Pt: 8.3 °C
Flash Pt: 97.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00069418
CAS Number: 345-18-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty