Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-formylpyridine 98%

Supplier: AMBEED, INC
Warning

A106881-1G A106881-5G A106881-100MG A106881-10G A106881-250MG A106881-1G A106881-5G
103797-094EA 13 USD InStock
103797-094 103822-852 77093-774 77093-776 77093-778 77309-714 77309-716
5-Chloro-2-formylpyridine 98%
5-Chloro-2-formylpyridine
Precaución: for Research Use Only


Formula: C₆H₄ClNO
Boiling Pt: 207.9 °C (760 mm Hg)
Melting Pt: 66…68 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD08277271
CAS Number: 31181-89-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty