Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-hydroxybenzonitrile 97%

Supplier: AMBEED, INC
Warning

A202202-5G A202202-1G A202202-5G A202202-100G A202202-10G A202202-250MG A202202-25G
103817-364EA 6 USD InStock
103817-364 103848-000 77086-096 77086-098 77152-222 77152-224 77152-226 77302-798
5-Chloro-2-hydroxybenzonitrile 97%
5-Chloro-2-hydroxybenzonitrile
Precaución: for Research Use Only


Formula: C₇H₄ClNO
Boiling Pt: 269.4 °C (760 mm Hg)
Melting Pt: 152…154 °C
Flash Pt: 116.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00219003
CAS Number: 13589-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty