Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-methyl-4-nitroaniline 98%

Supplier: AMBEED, INC
Warning

A295915-1G A295915-25G A295915-5G A295915-100G A295915-10G A295915-250MG A295915-500G A295915-1G
103814-074EA 10 USD InStock
103814-074 103835-208 76837-872 76837-874 77141-410 77141-412 77141-414 77141-416 77305-680
5-Chloro-2-methyl-4-nitroaniline 98%
5-Chloro-2-methyl-4-nitroaniline
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O₂
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00043921
CAS Number: 13852-51-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty