Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-methylbenzoic acid 97%

Supplier: AMBEED, INC
Warning

A241940-5G A241940-100G A241940-10G A241940-1G A241940-500G A241940-25G A241940-5G
103851-256EA 7 USD InStock
103851-256 76771-438 76771-440 76771-442 76771-444 77090-418 77304-236
5-Chloro-2-methylbenzoic acid 97%
5-Chloro-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₇ClO₂
Melting Pt: 168 °C
Flash Pt: 131.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00045842
CAS Number: 7499-06-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty