Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-methylpyrazolo[1,5-a]pyrimidine 98%

Supplier: AMBEED, INC
Warning

A196349-1G A196349-250MG A196349-100MG
103816-836EA 128 USD InStock
103816-836 103847-448 77671-746 77881-446
5-Chloro-2-methylpyrazolo[1,5-a]pyrimidine 98%
5-Chloro-2-methylpyrazolo[1,5-a]pyrimidine
Precaución: Inert atmosphere, 2…8 °C.


Formula: C₇H₆ClN₃
Storage Temperature: Refrigerator
MDL Number: MFCD17676711
CAS Number: 189116-36-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty