Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A270722-1G A270722-5G A270722-1G A270722-25G A270722-100G A270722-10G
103813-010EA 19 USD InStock
103813-010 103852-916 77092-682 77092-684 77113-900 77113-902
5-Chloro-2-methylpyridine 98%
5-Chloro-2-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₆ClN
MW: 127.572 g/mol
Boiling Pt: 163 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD04114188
CAS Number: 72093-07-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty