Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-nitroaniline 98%

Supplier: AMBEED, INC
Danger

A507011-25G A507011-100G A507011-500G
103810-408EA 232 USD InStock
103810-408 103839-442 76843-424 77292-138 77292-140
5-Chloro-2-nitroaniline 98%
5-Chloro-2-nitroaniline
Precaución: for Research Use Only


Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Boiling Pt: 323
Melting Pt: 125…129 °C
Flash Pt: 149.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00007776
CAS Number: 1635-61-6
UN: 2237
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty