Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-nitrobenzoic acid 98%

Supplier: AMBEED, INC
Danger

A146727-5G A146727-10G A146727-5G A146727-25G
103841-788EA 5 USD InStock
103841-788 77272-824 77299-542 77317-126
5-Chloro-2-nitrobenzoic acid 98%
5-Chloro-2-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 362 °C (760 mm Hg)
Melting Pt: 137…139 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007290
CAS Number: 2516-95-2
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty