Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloropyrazine-2-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A102532-1G A102532-5G A102532-100MG A102532-250MG A102532-1G A102532-5G
103796-382EA 49 USD InStock
103796-382 103822-078 76821-288 76821-290 77308-956 77308-958
5-Chloropyrazine-2-carbaldehyde 97%
5-Chloropyrazine-2-carbaldehyde
Precaución: for Research Use Only


Formula: C₅H₃ClN₂O
MW: 142.54 g/mol
Boiling Pt: 211 °C (760 mm Hg)
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697774
CAS Number: 88625-24-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty