Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloropyrazine-2-carbonitrile 97%

Supplier: AMBEED, INC
Warning

A125934-1G A125934-10G A125934-250MG A125934-1G A125934-25G A125934-5G
103800-740EA 19 USD InStock
103800-740 76782-010 76782-012 77298-102 77312-904 77312-906
5-Chloropyrazine-2-carbonitrile 97%
5-Chloropyrazine-2-carbonitrile
Precaución: for Research Use Only


Formula: C5H2ClN3
Boiling Pt: 241.552 °C (760 mm Hg)
Melting Pt: 44…45 °C
Flash Pt: 99.887 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09607700
CAS Number: 36070-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty