Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloropyridine-3,4-diamine 97%

Supplier: AMBEED, INC
Warning

A157442-1G A157442-5G A157442-250MG A157442-1G A157442-25G
103805-008EA 54 USD InStock
103805-008 103843-208 76783-688 77319-030 77319-032
5-Chloropyridine-3,4-diamine 97%
5-Chloropyridine-3,4-diamine
Precaución: for Research Use Only


Formula: C5H6ClN3
MW: 143.57 g/mol
Boiling Pt: 359.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09263519
CAS Number: 929074-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty