Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Ethynyl-2-fluoropyridine 97%

Supplier: AMBEED, INC
Warning

A416720-1G A416720-5G A416720-250MG A416720-1G A416720-5G
103827-322EA 74 USD InStock
103827-322 103838-368 76829-402 77307-180 77307-182
5-Ethynyl-2-fluoropyridine 97%
5-Ethynyl-2-fluoropyridine
Precaución: for Research Use Only


Formula: C₇H₄FN
Storage Temperature: Refrigerator
MDL Number: MFCD13175289
CAS Number: 853909-08-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty