Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Ethynylbenzo[d][1,3]dioxole 97%

Supplier: AMBEED, INC
Warning

A100396-1G A100396-1MG A100396-250MG A100396-5G
103795-418EA 493 USD InStock
103795-418 76765-106 77623-804 77876-778
5-Ethynylbenzo[d][1,3]dioxole 97%
5-Ethynylbenzo[d][1,3]dioxole
Precaución: for Research Use Only


Formula: C₉H₆O₂
Boiling Pt: 222.9 °C (760 mm Hg)
Melting Pt: 31 °C
Flash Pt: 90.1 °C
Storage Temperature: Freezer
MDL Number: MFCD07777106
CAS Number: 57134-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty